Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JSRP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560811
Description
Immunogen sequence: EAEVRPKIPG SREAAENDEE EPGEATGEAV REDRVTLADR GPKERPRREG KPRKEKPRKE ERPKKERPRK EER Highest antigen sequence identity to the following orthologs: Mouse - 42%, Rat - 48%.
The junction between the transverse tubules (T-tubules) and the sarcoplasmic reticulum (SR) of skeletal muscle is called the triad. At the triad, dihydropyridine receptors (DHPR) of the T-tubule serve as voltage sensors in excitation-contraction coupling, while ryanodine receptors (RyR), the calcium release channels, exist in the membrane of the terminal cisternae of the SR. It is thought that during slow phase depolarization of the T-tubule, a third protein, triadin (95 kDa) transmits electrochemical signals to the SR through direct interaction with both DHPR and RyR. Another skeletal muscle specific 90 kDa protein has also been localized to the this junction. This protein is phosphorylated by an intrinsic protein kinase in triads suggesting that it may play an important regulatory or structural role in the skeletal muscle triad junction.
Specifications
JSRP1 | |
Polyclonal | |
Unconjugated | |
JSRP1 | |
2300003C06Rik; 2310032K21Rik; homolog of mouse skeletal muscle sarcoplasmic reticulum protein JP-45; JP45; JP-45; Jsrp1; Junctional sarcoplasmic reticulum protein 1; Junctional SR; junctional-face membrane protein of 45 kDa homolog; skeletal muscle sarcoplasmic reticulum protein JP-45 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
126306 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96MG2 | |
JSRP1 | |
Recombinant protein corresponding to Human JSRP1. Recombinant protein control fragment (Product #RP-99141). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction