Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JT010
GREENER_CHOICESDP

Catalog No. 626925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-692-5 25 mg
62-695 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-692-5 Supplier Tocris Bioscience™ Supplier No. 6269/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective TRPA1 channel activator (EC50 = 65 nM)

JT010 is a potent and selective TRPA1 channel activator (EC50 = 0.65 nM); activates the TRPA1 channel by covalently and site-selectively binding to Cys621. Exhibits selectivity for TRPA1 over TRPV1, TRPV3, TRPV4, TRPM2, TRPM8, and TRPC5 channels (EC50 values are >1 μM).

Specifications

Chemical Name or Material 2-Chloro-N-[4-(4-methoxyphenyl)-2-thiazolyl]-N-(3-methoxypropyl)acetamide
CAS 917562-33-5
Quantity 25 mg
Molecular Formula C16H19ClN2O3S
Formula Weight 354.85
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.