Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JTB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587891 Supplier Invitrogen™ Supplier No. PA587891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAPVQEEKLS ASTSNLPCWL VEEFVVAEEC SPCSNFRAKT TPECGPTGYV EKITCSSSKR NEFKSCRSAL MEQRL; Positive Samples: MCF7, NCI-H460, A375, Rat spleen, Rat thymus; Cellular Location: Cytoplasm, Membrane, Mitochondrion, Single-pass type I membrane protein, centrosome, cytoskeleton, microtubule organizing center, spindle.

Required for normal cytokinesis during mitosis. Plays a role in the regulation of cell proliferation. May be a component of the chromosomal passenger complex (CPC), a complex that acts as a key regulator of mitosis. The CPC complex has essential functions at the centromere in ensuring correct chromosome alignment and segregation and is required for chromatin-induced microtubule stabilization and spindle assembly. Increases AURKB activity. Inhibits apoptosis induced by TGFB1. Overexpression induces swelling of mitochondria and reduces mitochondrial membrane potential.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JTB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene JTB
Gene Accession No. O76095, O88823
Gene Alias hJT; HJTB; HSPC222; Jtb; jumping translocation breakpoint; jumping translocation breakpoint protein; PAR; PAR protein; Prostate androgen-regulated protein; protein JTB
Gene Symbols JTB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 31-105 of human JTB (NP_0066851).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10899, 29439
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.