Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JTB Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304953100UL
Description
JTB Polyclonal specifically detects JTB in Human, Rat samples. It is validated for Western Blot.Specifications
JTB | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Western Blot | |
Unconjugated | |
hJT, HJTB, HSPC222, jumping translocation breakpoint, Jumping translocation breakpoint protein, PAR, PAR protein, Prostate androgen-regulated protein, protein JTB | |
Recombinant fusion protein containing a sequence corresponding to amino acids 31-105 of human JTB (NP_006685.1). | |
100 μL | |
Cancer | |
10899 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction