Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JTV-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531306
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531306 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531306 Supplier Invitrogen™ Supplier No. PA531306
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116, HepG2, HeLa, A549. Predicted reactivity: Mouse (88%), Rat (90%), Pig (84%), Sheep (81%), Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

JTV1 is probable core protein of the multisynthetase complex that serves as a template for the assembly of the supramolecular structure. This protein Mediates ubiquitination of FUBP1 and its degradation by the proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JTV-1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene AIMP2
Gene Accession No. Q13155, Q8R010
Gene Alias AA407136; AA407225; AIMP2; Aimp2(p38); Aminoacyl tRNA synthase complex-interacting multifunctional protein 2; aminoacyl tRNA synthetase complex interacting multifunctional protein 2; aminoacyl tRNA synthetase complex-interacting multifunctional protein 2; ARS interacting multifunctional protein 2; ARS-interacting multi-functional protein 2; JTV1; JTV-1; multisynthase complex auxiliary component p38; multisynthetase complex auxiliary component p38; P38; PRO0992; Protein JTV-1
Gene Symbols AIMP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human AIMP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231872, 7965
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.