Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ JTV 519 fumarate
SDP

Catalog No. 456450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-645-0 50 mg
45-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-645-0 Supplier R&D Systems™ Supplier No. 4564/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Ryanodine receptor (RyR) inhibitor

JTV 519 fumarate is a ryanodine receptor (RyR) inhibitor; stabilizes RyR2 in a closed conformation. Exhibits antiarrhythmic and cardioprotective properties in vivo.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type JTV 519 fumarate
Purity 98%
Molecular Weight (g/mol) M.W. (Observed): 540.67

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.