Learn More
Julolidine, 98%
Description
Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Julolidine used as amine building block and in chemoluminescent dye. Also used in photoconductive materials, chemiluminescence substances, chromogenic substrates in analytical redox reactions, dye intermediates, potential antidepressants and tranquilizers, nonlinear optical materials, high sensitivity photopolymerizable materials, and for improving color stability in photography.
Solubility
Soluble in toluene. Insoluble in water.
Notes
Air sensitive. Store away from air, oxidizing agents, carbon dioxide. Store under dry inert gas.
Specifications
Specifications
| Melting Point | 32°C to 37°C |
| Boiling Point | 170°C to 173°C (31 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 1 g |
| Beilstein | 139925 |
| Sensitivity | Air sensitive |
| Solubility Information | Soluble in toluene. Insoluble in water. |
| Formula Weight | 173.26 |
| Percent Purity | 98% |
| Chemical Name or Material | Julolidine |
Safety and Handling
| EINECSNumber | 207-535-0 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.