Learn More
Trajan™ Jumbo Syringe
SDP
Catalog No. SG009910 Shop All SGE Inc. Products
Change view
Click to view available options
Volume (Metric):
500 mL
Catalog No. Volume (Metric)
SG009910 500 mL
1 options
Catalog No. SG009910 Supplier Trajan™ Supplier No. 009910
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
  • Designed for holding and dispensing large volumes of gas
  • Heavy duty acrylic barrels
  • Easy access to sample for the addition of standards or removal of subsample via secondary port
  • Plunger stem can be unscrewed for ease of transportation and storage
  • Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Material Acrylic, Acrylic (Barrel)
For Use With (Application) For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling
Product Type Jumbo Syringe
Model 500MAR-LL-GT
Volume (Metric) 500 mL