Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JunB/AP-1 Antibody (PCRP-JUNB-3G2), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308391AF647
Description
JunB/AP-1 Monoclonal antibody specifically detects JunB/AP-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
JunB/AP-1 | |
Monoclonal | |
Alexa Fluor 647 | |
activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B | |
Recombinant full-length human JunB/AP-1 protein (Uniprot: P17275) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-JUNB-3G2 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
3726 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction