Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

JunB Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501994047
Encompass
Change view
Click to view available options
Quantity:
10 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4047 10 μL
50-199-4046 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4047 Supplier Active Motif Supplier No. 39550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

JunB ,a member of the Jun cellular oncogene family, is a basic region, leucine zipper transcription factor. It is involved in regulating gene activity following the primary growth factor response. JunB binds to the DNA sequence 5'-TGA[CG]TCA-3'. Cellular oncogenes play critical roles in cellular communication pathways that regulate growth, development, and differentiation.

Specifications

Antigen JunB
Applications Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene Junb
Gene Accession No. P17275
Gene Alias activator protein 1; AP-1; Jun B; jun B proto-oncogene; JUNB; Jun-b; Jun-B oncogene; JunB proto-oncogene, AP-1 transcription factor subunit; MyD21; Oncogene Jun-B; Transcription factor jun-B
Gene Symbols Junb
Host Species Rabbit
Immunogen This JunB antibody was raised against a synthetic peptide corresponding to amino acids 28-42 of human JunB.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3726
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.