Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Junctophilin-2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980454
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0454 Supplier Bioss Supplier No. BS0702R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein with a leucine-rich repeat and a calponin homology domain. Polymorphism in this gene may be associated with susceptibililty to knee osteoarthritis. Alternative splicing results in multiple transcript variants encoding different isoforms.

Specifications

Antigen Junctophilin-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene JPH2
Gene Accession No. Q9BR39
Gene Alias 1110002E14Rik; CMH17; JP2; JP-2; JP2NT; JPH2; jph2 {ECO:0000312; junctophilin 2; Junctophilin type 2; Junctophilin-2; Junctophilin-2 N-terminal fragment; RGD:1305196}
Gene Symbols JPH2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Junctophilin-2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57158
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.