Learn More
Invitrogen™ JunD Polyclonal Antibody

Description
Additional Information: This antibody is expected NOT to cross-react with Jun oncogen (NP_002219.1, GeneID 3725). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
Specifications
Specifications
| Antigen | JunD |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | JUND |
| Gene Accession No. | P15066 |
| Gene Alias | activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D |
| Gene Symbols | JUND |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.