Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JunD Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
Supplier: Active Motif 61403
Description
JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.Specifications
| JunD | |
| Polyclonal | |
| Unconjugated | |
| JUND | |
| activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; JUND; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3727 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP sequencing (ChIP-seq), Immunoprecipitation, Western Blot | |
| Lot-Specific | |
| PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
| P17535 | |
| JUND | |
| This antibody was raised against a peptide within the N-terminal region of human JunD. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction