Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JunD Rabbit anti-Human, Mouse, Rat, Clone: 5L7Y3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | JunD |
---|---|
Clone | 5L7Y3 |
Applications | ChIP Assay, Immunoprecipitation |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
JunD Monoclonal antibody specifically detects JunD in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, ImmunoprecipitationSpecifications
JunD | |
ChIP Assay, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human JunD (P17535). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5L7Y3 | |
Monoclonal | |
Purified | |
RUO | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3727 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title