Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JunD Recombinant Rabbit Monoclonal Antibody (SD0830)
GREENER_CHOICE

Catalog No. PIMA545022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545022 Supplier Invitrogen™ Supplier No. MA545022
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

JunD Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

JunD is a transcription factor which is coded by JunD gene. JunD transcriptional activity is modulated by phosphorylation in response to cellular stress via the c-Jun N terminal kinase (JNK)/Stress-Activated Protein Kinase (SAPK) family of protein kinase. Jun proteins are capable of forming dimers with Fos, ATF and CREB family transcription factors to form the AP-1 complex that differentially regulates a variety of target genes involved in cellular growth, proliferation, differentiation and apoptosis. Also, JunD plays an important role in metabolism via modulation of IGF-1 signaling pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JunD
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone SD0830
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene JUND
Gene Accession No. P17535
Gene Alias activator protein 1; AP1; AP-1; JUN D; jun D proto-oncogene; Jun proto-oncogene related gene d; Jun proto-oncogene related gene d1; Jund; Jun-d; JunD proto-oncogene, AP-1 transcription factor subunit; Jund1; JunD-FL isoform; Oncogene Jun-D; Transcription factor jun-D
Gene Symbols JUND
Host Species Rabbit
Immunogen Recombinant protein within Human JunD aa 101-200/347.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3727
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.