Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Justrite™ Base Cover for 30-Minute EN Cabinet
Shop All Justrite Safety Group Products
Click to view available options
For Use With (Equipment):
For 30 gal. EN cabinet
For 45 gal. EN cabinet
Description
- Made from melamine resin laminated chipboard
- Conceals integrated transport base on cabinet
- Prevents dust and debris from collecting underneath cabinet
- Creates seamless flush-to-floor attractive presentation
- Yellow
Specifications
Specifications
Description | Prevents collection of dust, debris; Melamine resin laminated chipboard |
Color | Yellow |
For Use With (Equipment) | For 30 gal. EN cabinet |
Material | Melamine Resin, Chipboard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction