Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Dispensing Safety Cans
GREENER_CHOICE

Catalog No. p-4367688 Shop All Justrite Safety Group Products
Click to view available options
Product Type:
Type I Shelf Safety Can
Type I Tilt Safety Can
Height (English):
15.8 in.
16.8 in.
Height (Metric):
403 mm
429 mm
Includes:
Tap 08902
Tap 08902 and Stand

Reliable dispensing features at an economical price

For safely dispensing hazardous liquids into test tubes, small flasks and beakers

  • Vacuum-relief valve within the lid for smooth liquid flow and self-closing faucets for trouble-free dispensing
  • Fill spouts have self-close, pressure-relief caps and stainless steel flame arresters
  • Capacity: 19L (5 gal)
  • Color: Red
  • Smaller 19-413-847 faucet offers easier dispensing in lab environments; No. 08902 faucet better suited for heavy, industrial use

Shelf Cans

  • Faucet mounted two inches above the bottom to eliminate lifting the cans during dispensing

Tilt Cans

  • Faucet on top with a sturdy metal cradle for the can; cradle base measures 35.1 x 35.6cm (13.81 x 14 in.)

Certifications

FM

Compliance

CARB, NFPA, OSHA

Specifications

Certifications/Compliance FM, CARB, NFPA, OSHA
Product Type Type I Shelf Safety Can
Closure Type Self Closing Pressure Relief Cap
Capacity (English) 5 gal.
Capacity (Metric) 19 L
Color Red
For Use With (Application) For Laboratories
Material Steel
Includes Tap 08902
Height (English) 16.8 in.
Height (Metric) 429 mm
Diameter (English) Inner 11.75 in.
Diameter (English) Opening 1.58 in.
Diameter (Metric) Inner 298 mm
Diameter (Metric) Opening 4 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.