Learn More
Justrite™ Biohazard Waste Can
Shop All Justrite Safety Group Products

Description
Safely segregate biohazard waste to ensure proper disposal of blood- or pathogen-contaminated laundry items and pathological and microbiological wastes.
- Features leakproof, sturdy steel construction with foot-operated, self-closing lid
- Eliminates any reason to reach into the waste area or touch the can
- Holds biohazard bag (sold separately) securely around circumference of can top
- Large biohazard label clearly identifies the contents and alerts workers
Not to be used for disposal of sharp items.
Certifications
FM approved
Compliance
Complies with OSHA 29CFR1910.1030.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Certifications/Compliance | FM, TUV, OSHA |
| Diameter (Metric) Outer | 35.56 cm |
| Diameter (English) Outer | 14 in. |
| Capacity (English) | 10 gal. |
| Color | White |
| Height (Metric) | 35.5 cm |
| Includes | Foot-operated Cover |
| For Use With (Application) | Safe, temporary collection of biological waste |
| Height (English) | 18 in. |
| Material | Metal |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.