Learn More
Justrite™ Horizontal and Vertical Drum Vents
Catalog No. p-33620
Click to view available options
Includes:
Operating Valve
Operating Valve, Plated Steel Flame Arrester
Operating Valve, Stainless Steel Flame Arrester
Product Type:
Horizontal Drum Vent
Vertical Drum Vent
Material:
Brass
Non-metallic Cast Iron
Stainless Steel
For controlled release of flammable vapors; prevent explosions caused by pressure buildup
  • Choose brass, stainless steel or nonmetallic vent
  • For petroleum-based liquids, use non-metallic or brass vents
  • For chlorinated solvents, use nonmetallic vent with stainless-steel flame arrester
  • For corrosives and general solvents, use stainless steel vents
Vertical Drum Vent:
  • Has operating valve that opens automatically to prevent pressure buildup inside drum
Horizontal Drum Vent
  • Has operating valve that can be opened manually to relieve internal pressure
Order Info
All vents fit 2 in. (5.1cm) bung. Cast Iron EL Fitting for use with 17-186A and 17-186B also available; see cat. no. 19018555.
Certifications
FM approved.
Specifications
Certifications/Compliance FM
Product Type Vertical Drum Vent
Description Opens at 5 psig Internal Pressure
For Use With (Application) Controlled release of flammable vapors (Petroleum-based liquids), prevent explosions caused by pressure buildup
For Use With (Equipment) Vertical Drums
Material Brass
Includes Operating Valve
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.