Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Sure-Grip™ EX Piggyback Hazardous Material Steel Safety Cabinet
GREENER_CHOICE

Catalog No. 19414805 Shop All Justrite Safety Group Products
Click to view available options
Dimensions (D x W x H) Interior:
14.5 x 39.5 x 13 in.
14.5 x 39.5 x 19.5 in.

Add another level of security to your work environment. The Justrite Sure-Grip™ EX Piggyback Hazardous Material Steel Safety Cabinet segregates hazardous materials using cabinets with an exclusive label package that ensures full regulatory compliance.

  • Designed to mount on top of standard 114L and 170L safety cabinets
  • Used independently as wall-mount unit or on countertop
  • Features double-wall construction, dual vents, grounding wire connections, adjustable shelves, leakproof sills, three-point self-latching doors and leveling feet
  • Automatic self-close door(s) shut and latch when fusible link melts at 74°C (165°F) under fire conditions
  • Polyethylene trays to mount on top of galvanized steel shelves
  • Separated polyethylene liner for bottom sump resists aggressive chemicals
  • Epoxy coated baked on powder-coat finish on both interior and exterior surface ensures chemical resistance
  • Each cabinet includes Hazardous - Keep Fire Away label and pack of ten application-specific labels to indicate its contents

Compliance

NFPA, U (NFPA)

Warranty and Services

10 Year

Specifications

Dimensions (D x W x H) Interior 14.5 x 39.5 x 13 in.
Product Type Combi Safety Storage Cabinet
Door Style Self-close
Load Bearing Capacity 12 gal. (45 L)
For Use With (Application) Hazardous materials
No. of Doors 2
Color Blue
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.