Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ JW 480
GREENER_CHOICESDP

Catalog No. 432750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-275-0 50 mg
43-271-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-275-0 Supplier R&D Systems™ Supplier No. 4327/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective inhibitor of serine hydrolase KIAA1363 (AADACL1)

Potent and selective inhibitor of the serine hydrolase enzyme KIAA1363 (AADACL1) (IC50 = 20 nM in mouse brain). Impairs migration, invasion, survival and tumor growth of prostate cancer cell lines in vivo. Reduces monoalkylglycerol ether (MAGE) levels.

Chemical Identifiers

CAS 1354359-53-7
Molecular Formula C22H23NO2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 333.42
Chemical Name or Material JW 480
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.