Learn More
EAAT3 Antibody (S180-41), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259319G
Catalog No. NBP259319G
Only null left
Add to Cart
Description
Description
EAAT3 Monoclonal antibody specifically detects EAAT3 in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
EAAT3
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
SLC1A1
Fusion protein amino acids 1-524 (full length) of human EAAT3 (Uniprot: P43005)
100 μL
6505
Human, Rat
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
S180-41
Western Blot, Immunocytochemistry/Immunofluorescence
EAAC1excitatory amino acid carrier 1, EAAT3excitatory amino acid transporter 3, Excitatory amino-acid carrier 1, Neuronal and epithelial glutamate transporter, Sodium-dependent glutamate/aspartate transporter 3, solute carrier family 1 (neuronal/epithelial high affinity glutamatetransporter, system Xag), member 1, Solute carrier family 1 member 1
Mouse
Protein G purified
Primary
Detects 60kDa.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.