Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JWA Mouse anti-Human,Mouse,Rat, KHL, Clone: 967C3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABT150125UG
Description
JWA Monoclonal antibody specifically detects JWA in Human,Mouse,Rat samples. It is validated for Immunoprecipitation, Western Blotting.
Specifications
JWA | |
Monoclonal | |
KLH | |
PRA1 family protein 3;ADP-ribosylation factor-like protein 6-interacting protein 5;ARL-6-interacting protein 5;Aip-5;Cytoskeleton-related vitamin A-responsive protein;Dermal papilla-derived protein 11;GTRAP3-18;Glutamate transporter EAAC1-interacting protein;JM5;Prenylated Rab acceptor protein 2;Protein JWa;Putative MAPK-activating protein PM27 | |
Mouse | |
∼22 kDa observed; 21.62 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
25 μg | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunoprecipitation, Western Blot | |
967C3 | |
NP_006398 | |
ARL6IP5, DERP11, JWA, PRA2, PRAF3 | |
KLH-conjugated linear peptide corresponding to 13 amino acids from the N-terminal region of human PRA1 family protein 3 (JWA). | |
Protein G Purified | |
Apoptosis and Cancer | |
Clone 967C3 is a mouse monoclonal antibody that detects PRA1 family protein 3 (JWA). It targets an epitope with in 13 amino acids from the N-terminal region. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction