Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ JWA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571372
Description
JWA Polyclonal Antibody for Western Blot
JWA protein expression is affected by vitamin A and may be associated with the cytoskeleton. A protein similar to JWA in rats may play a role in the regulation of cell differentiation by inhibiting the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport.
Specifications
| JWA | |
| Polyclonal | |
| Unconjugated | |
| Arl6ip5 | |
| 5930404D22Rik; addicsin; addiscin; ADP ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor GTPase 6 interacting protein 5; ADP-ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor-like 6 interacting protein 5; ADP-ribosylation factor-like protein 6-interacting protein 5; ADP-ribosylation-like factor 6 interacting protein 5; Aip5; Aip-5; ARL-6-interacting protein 5; Arl6ip5; AV001879; cytoskeleton related vitamin A responsive protein; cytoskeleton-related vitamin A-responsive protein; dermal papilla derived protein 11; dermal papilla-derived protein 11; DERP11; glutamate transporter EAAC1 interacting protein; glutamate transporter EAAC1-interacting protein; glutamate transporter EEAC1-associated protein; GTRAP3-18; hp22; HSPC127; JM5; jmx; Jwa; PRA1 domain family 3; PRA1 family protein 3; Pra2; Praf3; Prenylated Rab acceptor protein 2; protein JWa; putative MAPK activating protein PM27; putative MAPK-activating protein PM27; Yip6b | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 65106 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8R5J9 | |
| Arl6ip5 | |
| synthetic peptide directed towards the N terminal region of human JWA. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction