Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ JWA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109412 Supplier Invitrogen™ Supplier No. PA5109412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TFVMVVMLAS YFLISMFGGV MVFVFGITFP LLLMFIHASL RLRNLKNKLE NKMEGIGLKR TPMGIVLDAL EQQEEGINRL TDYISKVKE.

JWA protein expression is affected by vitamin A and may be associated with the cytoskeleton. A protein similar to JWA in rats may play a role in the regulation of cell differentiation by inhibiting the cell membrane glutamate transporter EAAC1. The expression of the rat gene is upregulated by retinoic acid, which results in a specific reduction in EAAC1-mediated glutamate transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JWA
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Arl6ip5
Gene Accession No. Q8R5J9
Gene Alias 5930404D22Rik; addicsin; addiscin; ADP ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor GTPase 6 interacting protein 5; ADP-ribosylation factor like GTPase 6 interacting protein 5; ADP-ribosylation factor-like 6 interacting protein 5; ADP-ribosylation factor-like protein 6-interacting protein 5; ADP-ribosylation-like factor 6 interacting protein 5; Aip5; Aip-5; ARL-6-interacting protein 5; Arl6ip5; AV001879; cytoskeleton related vitamin A responsive protein; cytoskeleton-related vitamin A-responsive protein; dermal papilla derived protein 11; dermal papilla-derived protein 11; DERP11; glutamate transporter EAAC1 interacting protein; glutamate transporter EAAC1-interacting protein; glutamate transporter EEAC1-associated protein; GTRAP3-18; hp22; HSPC127; JM5; jmx; Jwa; PRA1 domain family 3; PRA1 family protein 3; Pra2; Praf3; Prenylated Rab acceptor protein 2; protein JWa; putative MAPK activating protein PM27; putative MAPK-activating protein PM27; Yip6b
Gene Symbols Arl6ip5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human ARL6IP5 (NP_006398.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65106
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.