Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
JWA Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$149.33 - $480.74
Specifications
| Antigen | JWA |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
JWA Polyclonal specifically detects JWA in Mouse samples. It is validated for Western Blot.Specifications
| JWA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 10550 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| ADP-ribosylation factor-like protein 6-interacting protein 5, ADP-ribosylation-like factor 6 interacting protein 5, Aip-5, ARL-6-interacting protein 5, Cytoskeleton-related vitamin A-responsive protein, Dermal papilla-derived protein 11, DERP11addicsin, Glutamate transporter EAAC1-interacting protein, glutamate transporter EEAC1-associated protein, GTRAP3-18aip-5, hp22, HSPC127, JM5, jmx, JWAcytoskeleton related vitamin A responsive protein, PRA1 domain family 3, PRA1 family protein 3, PRA2, PRAF3dermal papilla derived protein 11, Prenylated Rab acceptor protein 2, Protein JWa, putative MAPK activating protein PM27, Putative MAPK-activating protein PM27 | |
| A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human JWA (NP_006398.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title