Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ JX 06
GREENER_CHOICESDP

Catalog No. 584550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
584550 50 mg
584510 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 584550 Supplier Tocris Bioscience™ Supplier No. 5845/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor

JX 06 is a potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor (IC50 values are 49, 101 and 313 nM for PDK1, PDK2 and PDK3, respectively). Exhibits selectivity for PDK1/2/3 over PDK4 and a panel of 323 other kinases. Suppresses A549 lung cancer cell proliferation in vitro. Reduces tumor volume of A549 xenografts in mice.

Specifications

Chemical Name or Material Bis(morpholinothiocarbonyl) disulfide
CAS 729-46-4
Quantity 50 mg
Target PDK-1 Inhibitors
Molecular Formula C10H16N2O2S4
Formula Weight 324.51
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.