Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ K 02288
GSA_VAGREENER_CHOICE

Catalog No. 498650R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-865-0R 50 mg
49-861-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-865-0R Supplier Tocris Bioscience™ Supplier No. 4986/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Type 1 BMP receptor inhibitor

K 02288 is a potent and selective inhibitor of type I bone morphogenic protein (BMP) receptors (IC50 values are 1.1, 1.8, 6.4, 34.4, 220, 302 and 321 nM for ALK2, ALK1, ALK6, ALK3, ActRIIA, ALK4 and ALK5 respectively). Reduces BMP4-induced Smad1/5/8 phosphorylation in vitro (IC50 = 100 nM) and induces dorsalization of zebrafish embryos.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 50 mg
Product Type K 02288
Purity >99%
Molecular Weight (g/mol) 352.38
Inhibitors K 02288

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.