Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ K 03861
GREENER_CHOICESDP

Catalog No. 678950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-895-0 50 mg
67-891-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 67-895-0 Supplier Tocris Bioscience™ Supplier No. 6789/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent CDK2 inhibitor

K 03861 is a potent Cdk2 inhibitor (Kd values are 9.7, 15.4, 18.6 and 50 nM at Cdk2 (C118L/A144C), Cdk2 (A144C), Cdk2 (C118L), and Cdk (WT) respectively. Competes with cyclin binding to inhibit Cdk2 kinase activity in vitro.

Specifications

Chemical Name or Material N-[4-[(2-Amino-4-pyrimidinyl)oxy]phenyl]-N'-[4-[(4-methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]urea
CAS 853299-07-7
Quantity 50 mg
Target Cyclin-dependent Kinase Inhibitors
Molecular Formula C24H26F3N7O2
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.