Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K-252a, 98%, , Thermo Scientific Chemicals

Catalog No. AC328311000
Change view
Click to view available options
Quantity:
1 mg
100 μg
Catalog No. Quantity
AC328311000 100 μg
AC328310010 1 mg
2 options

Catalog No. AC328311000

Supplier: Thermo Scientific Chemicals 328311000

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97161-97-2
Molecular Formula C27H21N3O5
Molecular Weight (g/mol) 467.5
MDL Number MFCD00161522
InChI Key KOZFSFOOLUUIGY-UHFFFAOYSA-N
Synonym methyl 6-hydroxy-5-methyl-13-oxo-5,6,7,8,14,15-hexahydro-13h-5,8-epoxy-4b,8a,14-triazadibenzo b,h cycloocta 1,2,3,4-jkl cyclopenta e-as-indacene-6-carboxylate
PubChem CID 3813
SMILES CC12C(CC(O1)N3C4=CC=CC=C4C5=C6C(=C7C8=CC=CC=C8N2C7=C53)CNC6=O)(C(=O)OC)O

Specifications

Assay Percent Range 97.5% min. (HPLC)
Quantity 100 μg
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in dichloromethane,dmso
Formula Weight 467.5
Percent Purity 98%
Chemical Name or Material K-252a
RTECSNumber NZ0550000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.