Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K-cadherin Monoclonal Antibody (427909)
GREENER_CHOICE

Catalog No. PIMA524159
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524159 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524159 Supplier Thermo Scientific Supplier No. MA524159
Add to Cart
Add to Cart

K-cadherin Monoclonal Antibody (427909)

K-cadherin Monoclonal Antibody for Western Blot, ICC/IF, Flow

TRUSTED_SUSTAINABILITY

Specifications

Antigen K-cadherin
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 427909
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CDH6
Gene Accession No. P55285
Gene Alias CAD6; CADH6; cadherin 6; cadherin 6, type 2, K-cadherin; cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin6; cadherin-6; Cdh6; k cadherin; KCAD; K-cadherin; Kidney cadherin
Gene Symbols CDH6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cadherin-6/KCAD Ser54-Ala615.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1004
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.