Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ K-cadherin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547373
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547373 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547373 Supplier Invitrogen™ Supplier No. PA547373
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Cadherin-8, rhCadherin-11, and rhCadherin-12 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Cadherins constitute a family of transmembrane glycoproteins involved in Ca2+-dependent cell-cell interactions. The members of this family are differentially expressed in various tissues. They function in the maintenance of tissue integrity and morphogenesis. Cadherins are divided into type I and type II subgroups. Type I cadherins include epithelial cadherin (E-cadherin, cadherin-1 or uvomorulin), neural cadherin (N-cadherin or cadherin-2), placental cadherin (P-cadherin or cadherin-3) and retinal cadherin (R-cadherin or cadherin-4), whereas kidney cadherin (K-cadherin or cadherin-6) and osteoblast cadherin (OB-cadherin or cadherin-11) are type II cadherins. Cadherin-6 is expressed in the proximal tubule epithelium of the kidney from which most renal cell carcinomas are derived.
TRUSTED_SUSTAINABILITY

Specifications

Antigen K-cadherin
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CDH6
Gene Accession No. P55285
Gene Alias CAD6; CADH6; cadherin 6; cadherin 6, type 2, K-cadherin; cadherin 6, type 2, K-cadherin (fetal kidney); Cadherin6; cadherin-6; Cdh6; k cadherin; KCAD; K-cadherin; Kidney cadherin
Gene Symbols CDH6
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cadherin-6 Thr22-Ala615.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1004
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.