Learn More
K'NEX Education™ Forces, Energy and Motion Set

Description
Includes
- 442 K'NEX™ pieces
- Color-coded building instructions
- Two battery motors
- Two flywheel motors
- Four transparent spring motors
- Comprehensive teacher's guide with seven inquiry-based lessons
Powerful and dynamic tools for creative learning.
- Hands-on investigation of potential energy and motion, inertia, speed and friction
- Students will build and experiment with 11 models powered by flywheel, spring and battery motors and wind power
- Uses rubber band spring motor-powered vehicles to investigate potential and kinetic energy
- Builds 11 curriculum-related models; builds up to four models at a time
- Grades 5 to 9
Key Concepts Include:
- Potential Energy (Gravitational and Elastic)
- Speed/Velocity
- Kinetic Energy
- Friction
- Inertia
- Motion and Force
Specifications
Specifications
| Grade | 5 to 9 |
| For Use With (Application) | Designed for students to investigate a variety of concepts related to forces, energy, and motion |
| Includes | 442 k'nex™ pieces, color-coded building instructions, two battery motors, two flywheel motors, four transparent spring motors, comprehensive teacher's guide with seven inquiry-based lessons |
| Class Size | 12 to 16 Students |
| Product Type | Forces, Energy and Motion Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.