Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K'NEX Education™ Introduction to Structures: Bridges Set
SCI_ED

Catalog No. S67346
Change view
Click to view available options
Grade:
3 to 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S67346 3 to 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S67346 Supplier K'NEX Education™ Supplier No. 78640

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Includes

  • 207 K'NEX™ pieces
  • Building instructions
  • Teacher's guide includes lesson plans, key concepts, terms, definitions, objectives and reproducible student worksheets on CD, aligned to national and state standards

Construct, investigate and experiment with seven different types of bridges.

Students construct, investigate and experiment with seven types of bridges.

  • Builds 13 bridge models, one model at a time
  • Models demonstrate seven key bridge types: arch, beam, bascule, cantilever, cable-stayed, truss and suspensions
  • Grades 5 to 9

Key Concepts Include:

  • Design Process
  • Compression
  • Tension
  • Load (Dead and Live)
  • Force
  • Stress
  • Cantilever Construction
  • Suspension Technology

Specifications

Product Type Introduction to Bridges Construction Set
Grade 3 to 6
For Use With (Application) Models demonstrate 7 Key bridge types: beam, truss, arch, cantilever, suspension, cable stayed and movable/bascule
Includes 207 K'NEX™ pieces, building instructions, teachers guide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.