Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ K-Ras (G12A Mutant) Recombinant Rabbit Monoclonal Antibody (HL1153)
GREENER_CHOICE

Catalog No. PIMA546940
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546940 Supplier Invitrogen™ Supplier No. MA546940
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

This gene, a Kirsten ras oncogene homolog from the mammalian ras gene family, encodes a protein that is a member of the small GTPase superfamily A single amino acid substitution is responsible for an activating mutation The transforming protein that results is implicated in various malignancies, including lung adenocarcinoma, mucinous adenoma, ductal carcinoma of the pancreas and colorectal carcinoma Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen K-Ras (G12A Mutant)
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL1153
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Accession No. P29466, P48729, Q8N752
Gene Alias ALPS4; C-BAS/HAS; CFC2; C-HA-RAS1; C-H-RAS; c-Ki-ras; c-Ki-ras2; c-K-ras; CMNS; CTLO; HAMSV; HRas proto-oncogene, GTPase; HRAS1; H-RASIDX; Ki-Ras; KRAS; K-Ras; KRAS proto-oncogene, GTPase; KRAS1; KRAS2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; NCMS; N-ras; NRAS proto-oncogene, GTPase; NRAS1; NS; NS3; NS6; p21ras; RALD; RASH1; RASK2
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide surrounding mutant G12A of human K-Ras. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122011, 1452, 834
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.