Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ K-Ras Monoclonal Antibody (A8E4)
GREENER_CHOICE

Catalog No. FEMA550601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550601 Supplier Invitrogen™ Supplier No. MA550601
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Rat brain tissue lysate, Jurkat cell lysate, N2A cell lysate, PC-12 cell lysate, human kidney tissue lysate, Hela, NIH/3T3. Predicted band size: 21 kDa Subcellular Location: Cell membrane Cytoplasm.

K-RAS (GTPase KRas) belongs to the Ras oncogene family. Ras proteins bind GDP/GTP and possess intrinsic GTPase activity. It plays an important role in the regulation of cell proliferation and has a role in promoting oncogenic events by inducing transcriptional silencing of tumor suppressor genes in colorectal cancer cells in a ZNF304-dependent manner. Mutations in the gene can result in leukemia acute myelogenous, leukemia juvenile myelomonocytic, Noonan Syndrome, gastric cancer, and cardiofaciocutaneous syndrome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen K-Ras
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A8E4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KRAS
Gene Accession No. P01116, P08644, P32883
Gene Alias AI929937; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; CFC2; c-Ki-ras; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; c-K-ras; c-K-ras2 protein; fa04e08; fc14b12; fc23g10; fj89d12; GTPase KRas; GTPase KRas, N-terminally processed; hypothetical protein LOC445289; K RAS; ki-Ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral (Kras-2) oncogene homolog; Kirsten rat sarcoma viral oncogene; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); KRAS; K-ras; K-Ras 2; K-ras p21 protein; KRAS proto-oncogene, GTPase; KRAS1; Kras2; Kras-2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; NS; NS3; oncogene KRAS2; p21; p21 protein; p21B; p21ras; PR310 c-K-ras oncogene; RALD; ras; ras p21; RASK; RASK2; ras-like protein; transforming protein p21; Unknown; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog; wu:fa04e08; wu:fc14b12; wu:fc23g10; wu:fj89d12; xRAS2; zgc:85725
Gene Symbols KRAS
Host Species Mouse
Immunogen Recombinant protein within human K-Ras aa 2-186.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16653, 24525, 3845
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.