Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ K-Ras4A, Isoform-specific, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABC1442MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABC1442MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC1442MI Supplier MilliporeSigma™ Supplier No. ABC1442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GTPase KRas (UniProt P01116; also known as c-K-ras, c-Ki-ras, K-Ras 2, Ki-Ras) is encoded by the KRAS (also known as KRAS2, RASK2) gene (Gene ID 3845) in human. Ras proteins are small GTPases controlling cellular pathways that signal for growth, proliferation, and differentiation. Mammalian genomes harbor three Ras genes (HRAS, NRAS, and KRAS) that encode four GTPases, two of which (K-Ras4A and K-Ras4B) are transcribed from the KRAS gene via two alternative fourth exons. Ras proteins share nearly identical sequence homology in sequence encoded by exons 1-3 (a.a. 1-165), while diverge in their C-terminal 24-a.a. hypervariable region (HVR) encoded by exon 4. The first 165 amino acids form the catalytic G domain that mediates GDP/GTP binding and association with effectors, exchange factors, and GTPase-activating proteins (GAPs), while the C-terminal HVR is needed for Ras membrane localization, a process necessary for Ras function. In addition to the removal of initiator methionine (Met1), Ras proteins are posttranslationally modified by farnesylation of Cys186, followed by proteolytical cleavage their C-terminal end three amino acids (a.a. 187-189) and carboxyl methylation of the newly formed C-terminal prenylcysteine. H-Ras, N-Ras, and K-Ras4A are further palmitoylated on cysteine residue(s) near the C-terminus (Cys181/Cys184 in H-Ras, Cys181 in N- and K-Ras), while K-Ras4B lacks a palmitoylation site. Palmitoylation is required for targeting H- and N-Ras to the plasma membrane (PM), while K-Ras4A and K-Ras4B contain a series of lysines that constitute a polybasic region (PBR) capable of interacting with the inner leaflet of the PM through an electrostatic interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen K-Ras4A, Isoform-specific
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene KRAS, KRAS2, RASK2
Gene Accession No. P01116
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to a sequence near the C-terminus of human K-Ras4A.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_203524
Test Specificity This rabbit polyclonal antibody specifically detected recombinant K-Ras4A, but not K-Ras4B, H-Ras or N-Ras (Tsai, F.D., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(3):779-784) by targeting a C-terminal region sequence unique to spliced isoform K-Ras4A (isoform 2A) and not found in spliced isoform K-Ras4B (isoform 2B).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.