Learn More
Greiner Bio-One K2EDTA GelTubes
Description
- Interior is spray dried with 1.8mg of anhydrous EDTA per 1mL of blood which acts as a anticoagulant binding the calcium ions and interrupting the clotting cascade
- Clear as glass with equal or better performance
- Molded to be virtually unbreakable, providing a better level of safety than glass
- Tubes should be centrifuged within six hours following blood collection
- Nonscrew Safety Pull Caps can be removed with a gentle pull
Specifications
Specifications
| Closure Type | Non-ridged Pull Cap |
| Capacity (Metric) | 5 mL |
| Closure Color | White with Yellow Ring |
| Tube Size | 13 x 100 mm |