Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

K2B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987983
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7983 Supplier Bioss Supplier No. BS7751R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIF2B is the plus end-directed microtubule-dependent motor required for spindle assembly and chromosome movement. KIF2B has microtubule depolymerization activity.

Specifications

Antigen K2B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KIF2B
Gene Accession No. Q8N4N8
Gene Alias 1700063D03Rik; EMBL:AAH83841.1, ECO:0000312; Kif2b; kif2b {ECO:0000312; kinesin family member 2B; kinesin protein; Kinesin-like protein KIF2B; Klp10A-like; LOC100338593; LOW QUALITY PROTEIN: kinesin-like protein KIF2B; RGD:1359248}; testis tissue sperm-binding protein Li 82P
Gene Symbols KIF2B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KIF2B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84643
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.