Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KA1/GRIK4/Glutamate Receptor KA1 Antibody (SR1043), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | KA1/GRIK4/Glutamate Receptor KA1 |
---|---|
Clone | SR1043 |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
KA1/GRIK4/Glutamate Receptor KA1 Monoclonal antibody specifically detects KA1/GRIK4/Glutamate Receptor KA1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
KA1/GRIK4/Glutamate Receptor KA1 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EAA1, Excitatory amino acid receptor 1, glutamate receptor KA1, Glutamate receptor KA-1, glutamate receptor, ionotropic kainate 4, glutamate receptor, ionotropic, kainate 4, KA1GRIK | |
A synthesized peptide derived from human KA1/GRIK4/Glutamate Receptor KA1 (Uniprot #: Q16099) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1043 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neuronal Cell Markers, Neurotransmission | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2900 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title