Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KA2/GRIK5/Glutamate Receptor KA2 Antibody (4G5H8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | KA2/GRIK5/Glutamate Receptor KA2 |
---|---|
Clone | 4G5H8 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61804
![]() |
Novus Biologicals
NBP261804 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
KA2/GRIK5/Glutamate Receptor KA2 Monoclonal specifically detects KA2/GRIK5/Glutamate Receptor KA2 in Human samples. It is validated for Western Blot, ELISA.Specifications
KA2/GRIK5/Glutamate Receptor KA2 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
2901 | |
Purified recombinant fragment of human GRIK5 (AA: extra 21-166) expressed in E. Coli. | |
Primary | |
109.3 kDa |
4G5H8 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
EAA2, Excitatory amino acid receptor 2, glutamate receptor KA2, Glutamate receptor KA-2, glutamate receptor, ionotropic kainate 5, glutamate receptor, ionotropic, kainate 5, KA2GRIK2 | |
GRIK5 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title