Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ kaiC Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144442 Supplier Invitrogen™ Supplier No. PA5144442
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

kaiC Polyclonal Antibody for Western Blot, ELISA

Central component of the KaiABC oscillator complex, which constitutes the main circadian regulator in cyanobacteria. Complex composition changes during the circadian cycle to control KaiC phosphorylation. KaiA stimulates KaiC autophosphorylation, while KaiB sequesters KaiA, leading to KaiC autodephosphorylation. Clock output pathways impact the RpaA transcriptional regulator. KaiC enhances the autophosphorylation activity of SasA, which then transfers its phosphate group to RpaA to activate it. KaiB and KaiC together enhance the phospho-RpaA dephosphatase activity of CikA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen kaiC
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Circadian clock protein kinase KaiC; kaiC
Host Species Rabbit
Immunogen Recombinant Nostoc sp. (strain PCC 7120/SAG 25.82/UTEX 2576) Circadian clock protein kinase KaiC (1-270aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.