Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Kainic acid (synthetic)
GREENER_CHOICE

Catalog No. 706510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-651-0 10 mg
70-655-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 70-651-0 Supplier Tocris Bioscience™ Supplier No. 7065/10
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Kainate agonist; excitant and neurotoxin

Kainic acid, also known as kainate, is a selective agonist of kainate ionotropic glutamate receptors (EC50 = 0.6 μM - 7.4 μM) and a partial agonist at AMPA receptors (EC50 = 31 μM - 170 μM). Kainic acid is used to model epilepsy in vivo and to study the mechanisms of neurodegeneration and neurocytosis induced by excess stimulation by kainic acid. Kainate is shown to be involved in amyloidogenic processing of amyloid precursor protein and Aβ peptides in Alzheimer's. Synthetic version of Kainic acid (Cat. No. 0222)

Related compounds include the kainate receptor agonist Domoic acid (Cat. No. 0269) and the antagonist NBQX (Cat. No. 0373).

Specifications

Chemical Name or Material (2S,3S,4S)-Carboxy-4-(1-methylethenyl)-3-pyrrolidineacetic acid
CAS 487-79-6
Quantity 10 mg
Molecular Formula C10H15NO4
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.