Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kaiso Antibody (6F) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001478
Description
Kaiso Monoclonal specifically detects Kaiso in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Western Blot, Microarray.Specifications
Kaiso | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1-2 ug/ml, Microarray | |
KAISOkaiso transcription factor, transcriptional regulator Kaiso, WUGSC:H_DJ525N14.1, zinc finger and BTB domain containing 33, Zinc finger and BTB domain-containing protein 33, ZNF348kaiso, ZNF-kaiso | |
Mouse | |
Protein A or G purified | |
RUO | |
10009 | |
Human, Mouse, Rat, Canine, Chicken | |
Purified |
Western Blot, Microarray | |
6F | |
Unconjugated | |
Q8BN78 | |
ZBTB33 | |
Recombinant purified mouse Kaiso protein (amino acids 1-499). | |
0.1 mg | |
Primary | |
Cross-reacts with Human, Mouse, Rat, Chicken and Dog. Not yet tested in other species. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction