Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kaiso Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581890
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581890 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581890 Supplier Invitrogen™ Supplier No. PA581890
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAELGVPLSQ VKSISGTAQD GNTEPLPPDS GDKNLVIQKS KDEAQDNGAT IMPIITESFS LSAEDYEMKK IIVTDSDDDD DDVIFCSEIL PTKETLPSNN TVAQVQSNPG PVAISD Highest antigen sequence indentity to the following orthologs: Mouse - 81%, Rat - 81%.

This gene encodes a transcriptional regulator with bimodal DNA-binding specificity, which binds to methylated CGCG and also to the non-methylated consensusKAISO-binding site TCCTGCNA. The protein contains an N-terminal POZ/BTB domain and 3 C-terminal zinc finger motifs. It recruits the N-CoR repressor complex to promote histone deacetylation and the formation of repressive chromatin structures in target gene promoters. It may contribute to the repression of target genes of the Wnt signaling pathway, and may also activate transcription of a subset of target genes by the recruitment of catenin delta-2 (CTNND2). Its interaction with catenin delta-1 (CTNND1) inhibits binding to both methylated and non-methylated DNA. It also interacts directly with the nuclear import receptor Importin-alpha 2 (also known as karyopherin alpha2 or RAG cohort 1), which may mediate nuclear import of this protein. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kaiso
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZBTB33
Gene Accession No. Q86T24
Gene Alias AW260253; E130014G12Rik; Kaiso; kaiso transcription factor; RGD1566309; RP23-322E15.9; Transcriptional regulator Kaiso; WUGSC:H_DJ525N14.1; Zbtb33; zinc finger and BTB domain containing 33; zinc finger and BTB domain-containing protein 33; zinc finger transcription factor Kaiso; ZNF348; ZNF-kaiso
Gene Symbols ZBTB33
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZBTB33. Recombinant protein control fragment (Product #RP-91977).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10009
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.