Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kaiso Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501565468
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5468 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5468 Supplier Bethyl Laboratories, Inc Supplier No. A303558AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

Kaiso is a novel member of the BTB/POZ (Broad complex, Tramtrak, Bric a brac/Pox virus and zinc finger) zinc finger superfamily of transcription factors. Yeast 2-hybrid studies have shown that Kaiso interacts with catenin p120ctn. This interaction involves the Arm repeats of 1-7 of the p120ctn Armadillo domain and the carboxyl-terminal third of Kaiso. The significance of the Kaiso:p120 ctn interaction is still unknown.

Specifications

Antigen Kaiso
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene ZBTB33
Gene Accession No. Q86T24
Gene Alias RP23-322E15.9, AW260253, E130014G12Rik, Kaiso, Znf-kaiso
Gene Symbols ZBTB33
Host Species Rabbit
Immunogen Between 622 and 672
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10009
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.