Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kaiso Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | Kaiso |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-5465
![]() |
Bethyl Laboratories, Inc
A303557AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Kaiso is a novel member of the BTB/POZ (Broad complex, Tramtrak, Bric a brac/Pox virus and zinc finger) zinc finger superfamily of transcription factors. Yeast 2-hybrid studies have shown that Kaiso interacts with catenin p120ctn. This interaction involves the Arm repeats of 1-7 of the p120ctn Armadillo domain and the carboxyl-terminal third of Kaiso. The significance of the Kaiso:p120 ctn interaction is still unknown.Specifications
Kaiso | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
RP23-322E15.9, AW260253, E130014G12Rik, Kaiso, Znf-kaiso | |
ZBTB33 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q86T24 | |
10009 | |
Between 375 and 425 | |
Primary | |
4° C | |
ZBTB33 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title