Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KAL1 Monoclonal Antibody (OTI5E10)
GREENER_CHOICE

Catalog No. PIMA527048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527048 Supplier Invitrogen™ Supplier No. MA527048
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KAL1 Monoclonal Antibody for IHC (P)

KAL1 (Anosmin-1) an extracellular matrix-associated glycosylated protein, have been linked with Kallmann Syndrome (KS), an X-linked genetic disorder characterized by loss of smell caused by abnormal olfactory bulb development and delayed puberty caused by disrupted migration of the gonadotropin-releasing hormone neuron. Anosmin-1 has been shown to directly bind FGFR1 via its N-terminal cysteine-rich domain, whey-acidic protein-like domain, and its first FnIII repeat with the D2 and D3 ectodomains of FGFR1. It is thought that Anosmin-1 can modulate FGFR1 signaling and have opposing effects on the formation and activation of FGF2-FGFR1-heparing complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAL1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI5E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ANOS1
Gene Accession No. P23352
Gene Alias Adhesion molecule-like X-linked; ADMLX; ANOS1; anosmin; anosmin 1; Anosmin1; anosmin-1; HH1; HHA; KAL; KAL1; KALIG1; KALIG-1; Kallmann syndrome interval gene 1; kallmann syndrome protein; Kallmann syndrome-1 sequence (anosmin-1); KMS; WAP four-disulfide core domain 19; WFDC19
Gene Symbols ANOS1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 129-373 of KAL1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3730
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.