Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kalirin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599843 Supplier Invitrogen™ Supplier No. PA599843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KALRN.

HAP1 (huntingtin-associated protein 1) binds to huntingtin. Huntingtin is a protein that contains a polyglutamine region and when the number of glutamine repeats exceeds 35, the gene encodes a version of huntingtin that leads to Huntington's disease (HD). The ability of HAP1 to bind to huntingtin is enhanced by an expanded polyglutamine repeat region. HAP1 shows neuronal localization and moves with huntingtin in nerve fibers. HAP1 is primarily expressed in brain tissue, with greater expression in the olfactory bulb and brain stem. Mouse HAP1 is localized to membrane-bound organelles including large endosomes, tubulovesicular structures and budding vesicles in neurons. Duo, also designated huntingtin-associated protein interacting protein or HAPIP, binds Huntingtin-associated protein 1 (HAP1) and may have a role in vesicle trafficking and cytoskeletal function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kalirin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KALRN
Gene Accession No. A2CG49, O60229, P97924
Gene Alias 2210407G14Rik; ARHGEF24; AV235988; CHD5; CHDS5; cyclin B1; DUET; DUO; E530005C20Rik; FLJ12332; FLJ16443; FLJ18196; FLJ18623; Gm539; HAPIP; hCG_2022551; huntingtin-associated protein interacting protein (duo); huntingtin-associated protein-interacting protein; Kalirin; kalirin, RhoGEF kinase; kalrn; PAM COOH-terminal interactor protein 10; P-CIP10; Protein Duo; RhoGEF kinase; serine/threonine kinase with Dbl- and pleckstrin homology domain; serine/threonine kinase with Dbl- and pleckstrin homology domains; serine/threonine-protein kinase with Dbl- and pleckstrin homology domain; TRAD
Gene Symbols KALRN
Host Species Rabbit
Immunogen A synthesized peptide derived from human KALRN(Accession O60229), corresponding to amino acid residues F1657-C1707.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 545156, 84009, 8997
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.