Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kalirin Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518214
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
HAP1 (huntingtin-associated protein 1) binds to huntingtin. Huntingtin is a protein that contains a polyglutamine region and when the number of glutamine repeats exceeds 35, the gene encodes a version of huntingtin that leads to Huntington's disease (HD). The ability of HAP1 to bind to huntingtin is enhanced by an expanded polyglutamine repeat region. HAP1 shows neuronal localization and moves with huntingtin in nerve fibers. HAP1 is primarily expressed in brain tissue, with greater expression in the olfactory bulb and brain stem. Mouse HAP1 is localized to membrane-bound organelles including large endosomes, tubulovesicular structures and budding vesicles in neurons. Duo, also designated huntingtin-associated protein interacting protein or HAPIP, binds Huntingtin-associated protein 1 (HAP1) and may have a role in vesicle trafficking and cytoskeletal function.
Specifications
| Kalirin | |
| Polyclonal | |
| Unconjugated | |
| KALRN | |
| 2210407G14Rik; ARHGEF24; AV235988; CHD5; CHDS5; cyclin B1; DUET; DUO; E530005C20Rik; FLJ12332; FLJ16443; FLJ18196; FLJ18623; Gm539; HAPIP; hCG_2022551; huntingtin-associated protein interacting protein (duo); huntingtin-associated protein-interacting protein; Kalirin; kalirin, RhoGEF kinase; kalrn; PAM COOH-terminal interactor protein 10; P-CIP10; Protein Duo; RhoGEF kinase; serine/threonine kinase with Dbl- and pleckstrin homology domain; serine/threonine kinase with Dbl- and pleckstrin homology domains; serine/threonine-protein kinase with Dbl- and pleckstrin homology domain; TRAD | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 8997 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O60229 | |
| KALRN | |
| Synthetic peptide sequence (RWHLGPGDPFSTYV) corresponding to the C-terminus amino acids of KALRN (aa 1641-1654). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction